Product Details

SNP ID
rs1408560
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:24328086 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCTTCCTCCTGCCTTGTGTGGGT[A/G]GGAAATTCCTTTCTGCCTTGCAACA
Phenotype
MIM: 608317
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRHL3 PubMed Links

Gene Details

Gene
GRHL3
Gene Name
grainyhead like transcription factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195010.1 Intron NP_001181939.1
NM_021180.3 Intron NP_067003.2
NM_198173.2 Intron NP_937816.1
NM_198174.2 Intron NP_937817.3
XM_011541869.1 Intron XP_011540171.1
XM_011541870.2 Intron XP_011540172.1

View Full Product Details