Product Details

SNP ID
rs1048227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40748283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCTGTTGCATCTCAAATTAATT[A/G]CTTTGAGATTTCTCTTCCAGGGAGT
Phenotype
MIM: 616646
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRT25 PubMed Links

Gene Details

Gene
KRT25
Gene Name
keratin 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181534.3 1396 Silent Mutation AGC,AGT S449S NP_853512.1
XM_011524414.1 1396 Silent Mutation AGC,AGT S447S XP_011522716.1

View Full Product Details