Product Details
- SNP ID
-
rs1058206
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:121339305 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAGCACTGTTACTCTAAGTCTCA[A/G]AAAAGGTTTTCAGTCAGGATATGGA
- Phenotype
-
MIM: 137350
MIM: 133090
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GSN
PubMed Links
Gene Details
- Gene
- GSN
- Gene Name
- gelsolin
There are no transcripts associated with this gene.
- Gene
- STOM
- Gene Name
- stomatin
View Full Product Details