Product Details

SNP ID
rs840381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:34022256 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGTAAGTTTCTGAGGAAATGGGT[C/T]TGCCAGAGACACAAAACCAGACTCC
Phenotype
MIM: 612045
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1QTNF3 PubMed Links

Gene Details

Gene
C1QTNF3
Gene Name
C1q and tumor necrosis factor related protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030945.3 Intron NP_112207.1
NM_181435.5 Intron NP_852100.3
Gene
C1QTNF3-AMACR
Gene Name
C1QTNF3-AMACR readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details