Product Details

SNP ID
rs8232
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:131500635 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTCCTGTTGGAGTAAGTTCCATA[C/T]ATAAAAGCTCATCCAAAACTTTGTT
Phenotype
MIM: 605623 MIM: 602632
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MKLN1 PubMed Links

Gene Details

Gene
MKLN1
Gene Name
muskelin 1
There are no transcripts associated with this gene.

Gene
PODXL
Gene Name
podocalyxin like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018111.2 5611 UTR 3 NP_001018121.1
NM_005397.3 5611 UTR 3 NP_005388.2

View Full Product Details