Product Details
- SNP ID
-
rs1733860
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:131501236 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCATTTCCAGTAAGACATTTACATT[A/C]CTGTCCAGAGAAAAAACTTGGCCAT
- Phenotype
-
MIM: 605623
MIM: 602632
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MKLN1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75399004] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MKLN1
- Gene Name
- muskelin 1
There are no transcripts associated with this gene.
- Gene
- PODXL
- Gene Name
- podocalyxin like
View Full Product Details