Product Details

SNP ID
rs3829397
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.13:44022923 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GACCCTAATGGTCACCTAAATGTGA[A/G]AGGAGGCTAGGCACTGAGAATGCCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00284 PubMed Links

Gene Details

Gene
LINC00284
Gene Name
long intergenic non-protein coding RNA 284
There are no transcripts associated with this gene.

View Full Product Details