Product Details
- SNP ID
-
rs10865804
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:25600614 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATATCATGTCTTACAAACATATTTT[A/C]CAAAACATCTTCAAGGGCTTTCCAG
- Phenotype
-
MIM: 180220
MIM: 126431
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
RARB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs150363543] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RARB
- Gene Name
- retinoic acid receptor beta
There are no transcripts associated with this gene.
- Gene
- TOP2B
- Gene Name
- topoisomerase (DNA) II beta
View Full Product Details