Product Details
- SNP ID
-
rs1059381
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:62410387 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAAGATTGTGCTTGGAGCTTGGCG[A/G]GCAGGGGTGGGGGTCCGAGTGGTCC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CABLES2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78496768] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CABLES2
- Gene Name
- Cdk5 and Abl enzyme substrate 2
There are no transcripts associated with this gene.
- Gene
- RBBP8NL
- Gene Name
- RBBP8 N-terminal like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_080833.2 |
2683 |
UTR 3 |
|
|
NP_543023.2 |
View Full Product Details