Product Details

SNP ID
rs883193
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:44342954 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACGAAACGCCCAGGTGGAGGGAGT[A/G]AGGCTCCCGTTTAGAGGCCAGTCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPATS1 PubMed Links

Gene Details

Gene
SPATS1
Gene Name
spermatogenesis associated serine rich 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145026.3 Intron NP_659463.1

View Full Product Details