Product Details

SNP ID
rs74315294
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:53202427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAATAAACATACAAGCTACATTT[C/T]GGGTAGGTAGGCTGGGCTGTGGGTA
Phenotype
MIM: 600650
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPT2 PubMed Links

Gene Details

Gene
CPT2
Gene Name
carnitine palmitoyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000098.2 521 Missense Mutation TCG,TTG S113L NP_000089.1
XM_005270484.1 521 Missense Mutation TCG,TTG S113L XP_005270541.1

View Full Product Details