Product Details

SNP ID
rs900109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:71200914 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACTCTGATGGTTATGTGCATTGC[A/G]GGACAGAAAACTAGTTATGAAAGAT
Phenotype
MIM: 601653
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EYA1 PubMed Links

Gene Details

Gene
EYA1
Gene Name
EYA transcriptional coactivator and phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000503.5 2677 Intron NP_000494.2
NM_001288574.1 2677 Intron NP_001275503.1
NM_001288575.1 2677 Intron NP_001275504.1
NM_172058.3 2677 Intron NP_742055.1
NM_172059.3 2677 Intron NP_742056.1
NM_172060.3 2677 Intron NP_742057.1
XM_011517483.2 2677 Intron XP_011515785.1
XM_011517484.2 2677 Intron XP_011515786.2
XM_017013201.1 2677 Intron XP_016868690.1
XM_017013202.1 2677 Intron XP_016868691.1
XM_017013203.1 2677 Intron XP_016868692.1
XM_017013204.1 2677 Intron XP_016868693.1
XM_017013205.1 2677 UTR 3 XP_016868694.1
XM_017013206.1 2677 Intron XP_016868695.1
XM_017013207.1 2677 Intron XP_016868696.1
XM_017013208.1 2677 Intron XP_016868697.1
XM_017013209.1 2677 Intron XP_016868698.1
XM_017013210.1 2677 Intron XP_016868699.1
XM_017013211.1 2677 Intron XP_016868700.1
XM_017013212.1 2677 Intron XP_016868701.1
XM_017013213.1 2677 Intron XP_016868702.1

View Full Product Details