Product Details
- SNP ID
-
rs1654587
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:6414391 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCGGAGGGCGGAGGCGCTAGGGTC[A/G]TAGGGGAGCTGCCCTGTCTCCGGAG
- Phenotype
-
MIM: 603445
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KHSRP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114825932] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KHSRP
- Gene Name
- KH-type splicing regulatory protein
- Gene
- LOC390877
- Gene Name
- adenylate kinase isoenzyme 1-like
There are no transcripts associated with this gene.
- Gene
- MIR3940
- Gene Name
- microRNA 3940
There are no transcripts associated with this gene.
View Full Product Details