Product Details

SNP ID
hCV8683588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:76021949 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTTTTAAGGAGATCTTTTAGAC[A/C]TGTAAGAAGAGAAAGGGGATATAAA
Phenotype
MIM: 603086 MIM: 147310
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ART3 PubMed Links

Gene Details

Gene
ART3
Gene Name
ADP-ribosyltransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130016.2 Intron NP_001123488.1
NM_001130017.2 Intron NP_001123489.1
NM_001179.5 Intron NP_001170.2
Gene
CXCL10
Gene Name
C-X-C motif chemokine ligand 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001565.3 Intron NP_001556.2

View Full Product Details