Product Details

SNP ID
rs1802172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30743339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTCAAGAAGATGGAAGGATCTC[A/T]CGGATCTCATTCCTAATGGTCCGCC
Phenotype
MIM: 606998 MIM: 602996
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FLOT1 PubMed Links

Gene Details

Gene
FLOT1
Gene Name
flotillin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318875.1 1100 Intron NP_001305804.1
NM_005803.3 1100 Intron NP_005794.1
XM_005248780.3 1100 Intron XP_005248837.1
XM_006714947.3 1100 Intron XP_006715010.1
XM_017010157.1 1100 Intron XP_016865646.1
XM_017010158.1 1100 Intron XP_016865647.1
Gene
HCG20
Gene Name
HLA complex group 20 (non-protein coding)
There are no transcripts associated with this gene.

Gene
IER3
Gene Name
immediate early response 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003897.3 1100 UTR 3 NP_003888.2

View Full Product Details