Product Details
- SNP ID
-
rs1246073
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:231740799 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTAGAAAGGTAGAACAGAGAGAAC[A/T]GAGGGGAGGATGGCATCAATAAATT
- Phenotype
-
MIM: 602676
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PDE6D
PubMed Links
Gene Details
- Gene
- PDE6D
- Gene Name
- phosphodiesterase 6D
View Full Product Details