Product Details

SNP ID
rs1407712
Assay Type
Functionally tested
NCBI dbSNP Submissions
33
Location
Chr.1:116495177 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTCCCTCCTCCTGCTCCTTCCTC[C/T]GGCTTCCCATCTCTCTCCTGTTTGG
Phenotype
MIM: 153420
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD58 PubMed Links

Gene Details

Gene
CD58
Gene Name
CD58 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144822.1 10827 Intron NP_001138294.1
NM_001779.2 10827 Intron NP_001770.1
XM_017002869.1 10827 UTR 3 XP_016858358.1
Gene
LOC105378922
Gene Name
uncharacterized LOC105378922
There are no transcripts associated with this gene.

View Full Product Details