Product Details

SNP ID
rs1061177
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:116493307 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGGGGCCTTTCCTTTGTCTAAC[G/T]GGCAATATCCTTACATCCGGGACCT
Phenotype
MIM: 153420
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CD58 PubMed Links

Gene Details

Gene
CD58
Gene Name
CD58 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144822.1 12697 Intron NP_001138294.1
NM_001779.2 12697 Intron NP_001770.1
XM_017002869.1 12697 UTR 3 XP_016858358.1
Gene
LOC105378922
Gene Name
uncharacterized LOC105378922
There are no transcripts associated with this gene.

View Full Product Details