Product Details

SNP ID
rs1710138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120538788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAGGTCAGGTATTATGAGGCAG[A/T]TATTTTGAGGTTGGTGGTATGAAGC
Phenotype
MIM: 616359 MIM: 615998
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
COQ5 PubMed Links

Gene Details

Gene
COQ5
Gene Name
coenzyme Q5, methyltransferase
There are no transcripts associated with this gene.

Gene
RNF10
Gene Name
ring finger protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014868.4 Intron NP_055683.3
XM_005254013.2 Intron XP_005254070.1
XM_006719717.2 Intron XP_006719780.1
XM_017020282.1 Intron XP_016875771.1
XM_017020283.1 Intron XP_016875772.1

View Full Product Details