Product Details
- SNP ID
-
rs995030
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:88496894 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACATTTCTGGAGCCATGCAAATGGT[A/G]CATGCAGTCTGAGACACGTGCTTTC
- Phenotype
-
MIM: 184745
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KITLG
PubMed Links
Gene Details
- Gene
- KITLG
- Gene Name
- KIT ligand
View Full Product Details