Product Details

SNP ID
rs1509415
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:110213450 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGTGTTCAGGCCTTTGTGTAATCC[A/C]CTCCCCTTGAGTGTGGGTGTGACCT
Phenotype
MIM: 607100
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC00116 PubMed Links

Gene Details

Gene
LINC00116
Gene Name
long intergenic non-protein coding RNA 116
There are no transcripts associated with this gene.

Gene
NPHP1
Gene Name
nephrocystin 1
There are no transcripts associated with this gene.

View Full Product Details