Product Details

SNP ID
rs1804959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:32541064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAAATACTCATTGACTACCTTAA[A/T]AATATGTCAATACCGTATGGCTTTA
Phenotype
MIM: 609507
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TOPORS PubMed Links

Gene Details

Gene
TOPORS
Gene Name
TOP1 binding arginine/serine rich protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195622.1 3458 UTR 3 NP_001182551.1
NM_005802.4 3458 UTR 3 NP_005793.2

View Full Product Details