Product Details

SNP ID
rs855465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33676029 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACAAACTGAGGATTGCAAGTTCCG[A/C]CACTGAACATTGGAATAGTTTCAAA
Phenotype
MIM: 613531
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PTENP1 PubMed Links

Gene Details

Gene
PTENP1
Gene Name
phosphatase and tensin homolog pseudogene 1
There are no transcripts associated with this gene.

Gene
PTENP1-AS
Gene Name
PTENP1 antisense RNA
There are no transcripts associated with this gene.

View Full Product Details