Product Details

SNP ID
rs1765003
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:4155202 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAGATTGGGGAAACAGCTGATCC[A/G]GCCCTCTAGCTCCAGCTGTGACGTG
Phenotype
MIM: 615854
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMOX PubMed Links

Gene Details

Gene
SMOX
Gene Name
spermine oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270691.1 Intron NP_001257620.1
NM_175839.2 Intron NP_787033.1
NM_175840.2 Intron NP_787034.1
NM_175841.2 Intron NP_787035.1
NM_175842.2 Intron NP_787036.1
XM_011529261.2 Intron XP_011527563.1

View Full Product Details