Product Details

SNP ID
rs918908
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:170178516 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTTGAAGGAGTAGGATAAACACC[A/C]TCTTGAGTATTGCTTTTTACACAGA
Phenotype
MIM: 610040
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MYO3B PubMed Links

Gene Details

Gene
MYO3B
Gene Name
myosin IIIB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083615.3 Intron NP_001077084.2
NM_138995.4 Intron NP_620482.3
XM_006712299.3 Intron XP_006712362.1
XM_011510654.2 Intron XP_011508956.1
XM_011510655.2 Intron XP_011508957.1
XM_011510656.2 Intron XP_011508958.1
XM_011510657.2 Intron XP_011508959.1
XM_011510658.2 Intron XP_011508960.1

View Full Product Details