Product Details

SNP ID
rs1801823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33291805 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTTGAAGCCTTTGCTTGGCCCTT[A/G]CATGTTAGGATATGGCCAAGAATCA
Phenotype
MIM: 605660 MIM: 602306 MIM: 601962 MIM: 611440
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6834 PubMed Links

Gene Details

Gene
MIR6834
Gene Name
microRNA 6834
There are no transcripts associated with this gene.

Gene
MIR6873
Gene Name
microRNA 6873
There are no transcripts associated with this gene.

Gene
PFDN6
Gene Name
prefoldin subunit 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185181.2 2994 Intron NP_001172110.1
NM_001265595.1 2994 Intron NP_001252524.1
NM_001265596.1 2994 Intron NP_001252525.1
NM_014260.3 2994 Intron NP_055075.1
Gene
RGL2
Gene Name
ral guanine nucleotide dissociation stimulator like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243738.1 2994 UTR 3 NP_001230667.1
NM_004761.4 2994 UTR 3 NP_004752.1
XM_011514781.1 2994 UTR 3 XP_011513083.1
XM_011514785.2 2994 Intron XP_011513087.1
Gene
TAPBP
Gene Name
TAP binding protein (tapasin)
There are no transcripts associated with this gene.

Gene
WDR46
Gene Name
WD repeat domain 46
There are no transcripts associated with this gene.

View Full Product Details