Product Details

SNP ID
rs7825
Assay Type
Validated
NCBI dbSNP Submissions
72
Location
Chr.1:111414475 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
GACACCAGAGGGGCTTAGGCTTCTT[C/G]ATCCACAGCAGAGTTTTCTGGGATT
Phenotype
MIM: 603578
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OVGP1 PubMed Links

Gene Details

Gene
OVGP1
Gene Name
oviductal glycoprotein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002557.3 2082 Missense Mutation CAA,GAA Q676E NP_002548.3

View Full Product Details