Product Details

SNP ID
rs1570171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:100972002 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAATACAGACTGGCGTGGGCAGAGA[A/T]GTGCACCCCCTTGGAGAGGAATACC
Phenotype
MIM: 611848 MIM: 610348
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MIR608 PubMed Links

Gene Details

Gene
MIR608
Gene Name
microRNA 608
There are no transcripts associated with this gene.

Gene
MRPL43
Gene Name
mitochondrial ribosomal protein L43
There are no transcripts associated with this gene.

Gene
SEMA4G
Gene Name
semaphorin 4G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001203244.1 Intron NP_001190173.1
NM_017893.3 Intron NP_060363.2
XM_005270008.2 Intron XP_005270065.1
Gene
SLF2
Gene Name
SMC5-SMC6 complex localization factor 2
There are no transcripts associated with this gene.

View Full Product Details