Product Details

SNP ID
rs1659563
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40448514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTGTGGGCCTTGGTGTAGATGCC[A/G]GGATGGCTCACCTCTGTCCTCAATC
Phenotype
MIM: 613880
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BAHD1 PubMed Links

Gene Details

Gene
BAHD1
Gene Name
bromo adjacent homology domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301132.1 Intron NP_001288061.1
NM_014952.4 Intron NP_055767.3
XM_011521366.2 Intron XP_011519668.2
XM_011521367.2 Intron XP_011519669.2
XM_011521368.2 Intron XP_011519670.2
XM_011521369.2 Intron XP_011519671.1

View Full Product Details