Product Details

SNP ID
rs1048704
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:72942371 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGCACCCTCACACTGCTGTCTGG[A/G]AATGGAAGAAGGGCATCCCCAAGCC
Phenotype
MIM: 600034 MIM: 615572
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EMX1 PubMed Links

Gene Details

Gene
EMX1
Gene Name
empty spiracles homeobox 1
There are no transcripts associated with this gene.

Gene
SFXN5
Gene Name
sideroflexin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144579.2 3812 UTR 3 NP_653180.1
XM_005264648.1 3812 UTR 3 XP_005264705.1
XM_006712147.1 3812 UTR 3 XP_006712210.1
XM_011533186.2 3812 UTR 3 XP_011531488.1
XM_011533187.2 3812 UTR 3 XP_011531489.1
XM_011533188.2 3812 UTR 3 XP_011531490.1
XM_011533189.1 3812 UTR 3 XP_011531491.1
XM_011533190.2 3812 UTR 3 XP_011531492.1
XM_011533192.2 3812 UTR 3 XP_011531494.1
XM_011533194.2 3812 UTR 3 XP_011531496.1
XM_011533195.2 3812 UTR 3 XP_011531497.1
XM_011533196.2 3812 UTR 3 XP_011531498.1
XM_011533197.1 3812 UTR 3 XP_011531499.1
XM_017005340.1 3812 UTR 3 XP_016860829.1
XM_017005341.1 3812 UTR 3 XP_016860830.1
XM_017005342.1 3812 UTR 3 XP_016860831.1
XM_017005343.1 3812 UTR 3 XP_016860832.1
XM_017005344.1 3812 UTR 3 XP_016860833.1
XM_017005345.1 3812 UTR 3 XP_016860834.1
XM_017005346.1 3812 UTR 3 XP_016860835.1

View Full Product Details