Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001135146.1 | 2263 | Intron | NP_001128618.1 | ||
NM_001135147.1 | 2263 | UTR 3 | NP_001128619.1 | ||
NM_001135148.1 | 2263 | Intron | NP_001128620.1 | ||
NM_022154.5 | 2263 | Intron | NP_071437.3 | ||
XM_005263177.1 | 2263 | Intron | XP_005263234.1 | ||
XM_011532181.2 | 2263 | Intron | XP_011530483.1 | ||
XM_011532182.1 | 2263 | Intron | XP_011530484.1 | ||
XM_017008540.1 | 2263 | Intron | XP_016864029.1 | ||
XM_017008541.1 | 2263 | Intron | XP_016864030.1 | ||
XM_017008542.1 | 2263 | Intron | XP_016864031.1 |