Product Details

SNP ID
rs1564502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:37261165 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTACCATTGCAGGCTTTGGCAGGTG[C/T]GGTGGCCTGTAATCCCAGAATTTTG
Phenotype
MIM: 616880
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TBC1D22B PubMed Links

Gene Details

Gene
TBC1D22B
Gene Name
TBC1 domain family member 22B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017772.3 Intron NP_060242.2
XM_011514738.2 Intron XP_011513040.1
XM_011514739.1 Intron XP_011513041.1
Gene
TMEM217
Gene Name
transmembrane protein 217
There are no transcripts associated with this gene.

View Full Product Details