Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001111125.2 | 4267 | Intron | NP_001104595.1 | ||
NM_001243197.1 | 4267 | Intron | NP_001230126.1 | ||
NM_015075.1 | 4267 | Intron | NP_055890.1 | ||
XM_006724579.2 | 4267 | Intron | XP_006724642.1 | ||
XM_006724580.3 | 4267 | Intron | XP_006724643.1 | ||
XM_006724581.3 | 4267 | UTR 3 | XP_006724644.1 | ||
XM_006724582.3 | 4267 | Intron | XP_006724645.1 | ||
XM_006724583.3 | 4267 | Intron | XP_006724646.1 | ||
XM_006724584.2 | 4267 | Intron | XP_006724647.1 | ||
XM_011530773.2 | 4267 | Intron | XP_011529075.1 | ||
XM_011530774.2 | 4267 | Intron | XP_011529076.1 | ||
XM_011530776.1 | 4267 | Intron | XP_011529078.1 | ||
XM_011530777.1 | 4267 | Intron | XP_011529079.1 | ||
XM_017029359.1 | 4267 | Intron | XP_016884848.1 | ||
XM_017029360.1 | 4267 | Intron | XP_016884849.1 |