Product Details
- SNP ID
-
rs1850565
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:22412655 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AACAAAGACCACTTCCCAGTCCTCC[A/G]GGGCGACACTTCCTGGAGATCCGTG
- Phenotype
-
MIM: 605160
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ36000
PubMed Links
Gene Details
- Gene
- FLJ36000
- Gene Name
- uncharacterized FLJ36000
There are no transcripts associated with this gene.
View Full Product Details