Product Details

SNP ID
rs4796715
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41837519 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTGACTTGGATTTCCTTGAGTTC[A/G]GTGTCTAGGGAGCCAAGTGTTGGGC
Phenotype
MIM: 608778
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHL10 PubMed Links

Gene Details

Gene
KLHL10
Gene Name
kelch like family member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011524706.2 200 UTR 5 XP_011523008.1
Gene
NT5C3B
Gene Name
5'-nucleotidase, cytosolic IIIB
There are no transcripts associated with this gene.

View Full Product Details