Product Details

SNP ID
rs7821027
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143645848 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCAGGATAGGCTCCAGCTACCCAG[A/G]ACCGTGCATTAAAATAAGTGGGTAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF623 PubMed Links

Gene Details

Gene
ZNF623
Gene Name
zinc finger protein 623
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001082480.2 Intron NP_001075949.1
NM_001261843.1 Intron NP_001248772.1
NM_014789.3 Intron NP_055604.3
XM_006716708.3 Intron XP_006716771.1

View Full Product Details