Product Details

SNP ID
rs2607292
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:10372506 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATACTGCTTGGAAATAATAATATGT[C/T]AGAATTCCCTTTAGGATTATTTTTT
Phenotype
MIM: 613297
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MARCH6 PubMed Links

Gene Details

Gene
MARCH6
Gene Name
membrane associated ring-CH-type finger 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270660.1 Intron NP_001257589.1
NM_001270661.1 Intron NP_001257590.1
NM_005885.3 Intron NP_005876.2
XM_011513932.2 Intron XP_011512234.1
XM_011513934.1 Intron XP_011512236.1
XM_011513936.2 Intron XP_011512238.1
XM_011513937.2 Intron XP_011512239.1
XM_017008944.1 Intron XP_016864433.1

View Full Product Details