Product Details

SNP ID
rs2007397
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:41144265 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGAAACTGAGGTCCCAAGAAGCC[A/G]AGAAATGTTACTCAAAGTCCCACAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00323 PubMed Links

Gene Details

Gene
LINC00323
Gene Name
long intergenic non-protein coding RNA 323
There are no transcripts associated with this gene.

View Full Product Details