Product Details

SNP ID
rs1455434
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40751141 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGCCAATGGGAATCGCCATTTAG[G/T]GTGCTCCGCCCACCGGGTCGCGTAG
Phenotype
MIM: 182285
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C19orf54 PubMed Links

Gene Details

Gene
C19orf54
Gene Name
chromosome 19 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198476.3 288 Intron NP_940878.3
XM_005258776.4 288 Intron XP_005258833.1
XM_005258778.4 288 Intron XP_005258835.1
XM_006723152.2 288 Intron XP_006723215.1
XM_006723153.2 288 Intron XP_006723216.1
XM_011526774.2 288 Intron XP_011525076.1
XM_011526775.1 288 Intron XP_011525077.1
XM_011526776.2 288 Intron XP_011525078.1
XM_011526777.2 288 Intron XP_011525079.1
XM_011526778.2 288 Intron XP_011525080.1
XM_011526779.2 288 Intron XP_011525081.1
XM_011526782.2 288 Intron XP_011525084.1
XM_011526783.2 288 Intron XP_011525085.1
XM_011526784.2 288 Intron XP_011525086.1
XM_011526785.2 288 Intron XP_011525087.1
XM_011526786.2 288 Intron XP_011525088.1
XM_017026620.1 288 Intron XP_016882109.1
XM_017026621.1 288 Intron XP_016882110.1
XM_017026622.1 288 Intron XP_016882111.1
XM_017026623.1 288 Intron XP_016882112.1
XM_017026624.1 288 Intron XP_016882113.1
XM_017026625.1 288 Intron XP_016882114.1
XM_017026626.1 288 Intron XP_016882115.1
XM_017026627.1 288 Intron XP_016882116.1
Gene
SNRPA
Gene Name
small nuclear ribonucleoprotein polypeptide A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004596.4 288 UTR 5 NP_004587.1

View Full Product Details