Product Details
- SNP ID
-
rs1046650
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
54
- Location
-
Chr.1:156669208 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGGCCAGGCCTCTCAGCCAGAAACC[A/G]TATGTCAAGAGATCGTAAGTTAAGA
- Phenotype
-
MIM: 600347
MIM: 600915
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BCAN
PubMed Links
Gene Details
- Gene
- BCAN
- Gene Name
- brevican
There are no transcripts associated with this gene.
- Gene
- NES
- Gene Name
- nestin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006617.1 |
5113 |
UTR 3 |
|
|
NP_006608.1 |
View Full Product Details