Product Details
- SNP ID
-
rs948820
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:66501356 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCTGAGTAATCTAATACAAAGCAA[A/G]ACAAAGCCAGGGTCACTGGAAGCAG
- Phenotype
-
MIM: 603016
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDH19
PubMed Links
Gene Details
- Gene
- CDH19
- Gene Name
- cadherin 19
View Full Product Details