Product Details

SNP ID
rs2338019
Assay Type
Validated
NCBI dbSNP Submissions
53
Location
Chr.1:151984338 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
AAAAGTATAGCAGCAGCATTTCCCA[T/C]ATGAGCACTGCAAACTCCTACTCTA
Phenotype
MIM: 114085
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
S100A10 PubMed Links

Gene Details

Gene
S100A10
Gene Name
S100 calcium binding protein A10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002966.2 Intron NP_002957.1

View Full Product Details