Product Details

SNP ID
rs1500942
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:153117212 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGTGAAATAATAAGATGGGCTA[A/C]CCGTGACACTTACTCTTATGTGGTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SPRR2F PubMed Links

Gene Details

Gene
SPRR2F
Gene Name
small proline rich protein 2F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014450.2 Intron NP_001014450.1
XM_017002176.1 Intron XP_016857665.1

View Full Product Details