Product Details

SNP ID
rs7103648
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47440232 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGTGAGGACTGGACCCAGTACCC[A/G]TAAGGACCCTGTGTGTGCTCAGTGC
Phenotype
MIM: 601592
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAPSN PubMed Links

Gene Details

Gene
RAPSN
Gene Name
receptor associated protein of the synapse
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005055.4 Intron NP_005046.2
NM_032645.4 Intron NP_116034.2
XM_005253042.3 Intron XP_005253099.1
XM_005253043.3 Intron XP_005253100.1
XM_011520252.1 Intron XP_011518554.1

View Full Product Details