Product Details
- SNP ID
-
rs831464
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:20157171 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGACTGCTCGACGCAGCATGCCCC[G/T]CCGAGGCTTCCCGCGCGCGGCCAGC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DBX1
PubMed Links
Gene Details
- Gene
- DBX1
- Gene Name
- developing brain homeobox 1
View Full Product Details