Product Details
- SNP ID
-
rs11583897
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
34
- Location
-
Chr.1:26361690 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTGCCCCATCACCAGGGGAGTCCC[C/G]GCACTGCTCAGGCTCACTGCTCTTG
- Phenotype
-
MIM: 616775
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
AIM1L
PubMed Links
Gene Details
- Gene
- AIM1L
- Gene Name
- absent in melanoma 1-like
There are no transcripts associated with this gene.
- Gene
- ZNF683
- Gene Name
- zinc finger protein 683
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001114759.2 |
1793 |
Silent Mutation |
GCC,GCG |
A492A |
NP_001108231.1 |
NM_001307925.1 |
1793 |
Silent Mutation |
GCC,GCG |
A512A |
NP_001294854.1 |
NM_173574.3 |
1793 |
Silent Mutation |
GCC,GCG |
A492A |
NP_775845.2 |
XM_005245828.3 |
1793 |
Silent Mutation |
GCC,GCG |
A520A |
XP_005245885.1 |
XM_005245830.3 |
1793 |
Silent Mutation |
GCC,GCG |
A512A |
XP_005245887.1 |
XM_005245832.3 |
1793 |
Silent Mutation |
GCC,GCG |
A512A |
XP_005245889.1 |
XM_006710555.3 |
1793 |
Silent Mutation |
GCC,GCG |
A521A |
XP_006710618.1 |
XM_011541198.2 |
1793 |
Silent Mutation |
GCC,GCG |
A512A |
XP_011539500.1 |
XM_017000954.1 |
1793 |
Silent Mutation |
GCC,GCG |
A521A |
XP_016856443.1 |
XM_017000955.1 |
1793 |
Silent Mutation |
GCC,GCG |
A521A |
XP_016856444.1 |
XM_017000956.1 |
1793 |
Silent Mutation |
GCC,GCG |
A520A |
XP_016856445.1 |
XM_017000957.1 |
1793 |
Silent Mutation |
GCC,GCG |
A501A |
XP_016856446.1 |
XM_017000958.1 |
1793 |
UTR 3 |
|
|
XP_016856447.1 |
XM_017000959.1 |
1793 |
UTR 3 |
|
|
XP_016856448.1 |
View Full Product Details