Product Details

SNP ID
rs34485759
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67124372 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCAAATACAACATTCTTCCTTTA[A/G]TTTTTGCCAAGCATGATGGTACTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C16orf70 PubMed Links

Gene Details

Gene
C16orf70
Gene Name
chromosome 16 open reading frame 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320540.1 Intron NP_001307469.1
NM_001320541.1 Intron NP_001307470.1
NM_001320542.1 Intron NP_001307471.1
NM_001320543.1 Intron NP_001307472.1
NM_025187.4 Intron NP_079463.2
XM_006721284.3 Intron XP_006721347.1
XM_011523350.2 Intron XP_011521652.1
XM_017023729.1 Intron XP_016879218.1
XM_017023730.1 Intron XP_016879219.1
XM_017023731.1 Intron XP_016879220.1

View Full Product Details