Product Details

SNP ID
rs35201481
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:17922075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAATGTATTCTCTAAGGTGTGA[C/T]TGCAGTTCAGGCTTTTTTTTTTTTT
Phenotype
MIM: 609198
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAMTSL1 PubMed Links

Gene Details

Gene
ADAMTSL1
Gene Name
ADAMTS like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040272.5 Intron NP_001035362.3
NM_052866.4 Intron NP_443098.3
XM_011518063.2 Intron XP_011516365.1
XM_011518064.2 Intron XP_011516366.1
XM_011518067.1 Intron XP_011516369.1
XM_011518068.2 Intron XP_011516370.1
XM_011518070.2 Intron XP_011516372.1
XM_017015310.1 Intron XP_016870799.1
XM_017015311.1 Intron XP_016870800.1
XM_017015312.1 Intron XP_016870801.1
XM_017015313.1 Intron XP_016870802.1
XM_017015314.1 Intron XP_016870803.1

View Full Product Details