Product Details

SNP ID
rs11835812
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:105020556 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTACCTGTTCATGAAACTTACATT[A/C]TAATGGGAAAAGATCATTGATTAAT
Phenotype
MIM: 613584
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ALDH1L2 PubMed Links

Gene Details

Gene
ALDH1L2
Gene Name
aldehyde dehydrogenase 1 family member L2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034173.3 6326 UTR 3 NP_001029345.2
XM_011537986.2 6326 UTR 3 XP_011536288.1
XM_011537988.2 6326 Intron XP_011536290.1
XM_011537989.2 6326 Intron XP_011536291.1
XM_017018889.1 6326 UTR 3 XP_016874378.1
XM_017018890.1 6326 Intron XP_016874379.1

View Full Product Details