Product Details

SNP ID
rs12686878
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:23986503 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
ACTCCCAGGTTATTCTTGCTTAACT[C/T]TGTCAACTATGTTTTTTCCTCTGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL15 PubMed Links

Gene Details

Gene
KLHL15
Gene Name
kelch like family member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030624.2 3701 UTR 3 NP_085127.2
XM_006724518.1 3701 UTR 3 XP_006724581.1

View Full Product Details